1-Methyl-3-octylimidazolium tetrafluoroborate ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

M2732-25G M2732-5G
TCM2732-25GEA 213.3 CAD
TCM2732-25G TCM2732-5G
1-Methyl-3-octylimidazolium tetrafluoroborate ≥97.0% (by HPLC, total nitrogen)
1-Methyl-3-octylimidazolium tetrafluoroborate
Formula: C₁₂H₂₃BF₄N₂
MDL Number: MFCD03095434
CAS Number: 244193-52-0

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR