DL-Threonine ≥97.0% (by titrimetric analysis) (contains DL-Allothreonine)

Supplier: TCI America

T3105-100G T3105-25G
TCT3105-100GEA 169.06 CAD
TCT3105-100G TCT3105-25G
DL-Threonine ≥97.0% (by titrimetric analysis) (contains DL-Allothreonine)
DL-Threonine
Formula: C₄H₉NO₃
Melting Pt: 230°C
MDL Number: MFCD00063722
CAS Number: 80-68-2
EINECS: 201-300-6
Merck Index: 14,09380

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR