Methyl-3,4-dimethoxybenzoate ≥98.0% (by GC)
Supplier: TCI America
M2805-25G
M2805-5G
TCM2805-25GEA
148.52
CAD
TCM2805-25G
TCM2805-5G
Methyl-3,4-dimethoxybenzoate ≥98.0% (by GC)
Methyl 3,4-dimethoxybenzoate
Formula:
C₁₀H₁₂O₄ Boiling Pt: 283°C |
MDL Number:
MFCD00008430 CAS Number: 2150-38-1 EINECS: 218-424-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 59.0 to 63.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...