Methyl-4-methoxy-3-nitrobenzoate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-Methoxy-3-nitrobenzoic acid methyl ester, Methyl 4-methoxy-3-nitrobenzoate

M2873-1G M2873-5G
TCM2873-1GEA 86.9 CAD
TCM2873-1G TCM2873-5G
Methyl-4-methoxy-3-nitrobenzoate ≥98.0% (by GC)
Methyl 4-methoxy-3-nitrobenzoate
Formula: C₉H₉NO₅
MW: 211.17 g/mol
MDL Number: MFCD00024213
CAS Number: 40757-20-8

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 108.0 to 112.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR