1,3-Dibenzoyloxybenzene ≥98.0% (by GC)
Supplier: TCI America
D5207-25G
D5207-5G
76219-238EA
148.52
CAD
76219-238
76219-240
1,3-Dibenzoyloxybenzene ≥98.0% (by GC)
1,3-Dibenzoyloxybenzene
Formula:
C₂₀H₁₄O₄ |
MDL Number:
MFCD00016576 CAS Number: 94-01-9 EINECS: 202-294-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 116.0 to 120.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...