2-Chloro-4-methoxybenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
C3077-1G
C3077-5G
TCC3077-1GEA
53.72
CAD
TCC3077-1G
TCC3077-5G
2-Chloro-4-methoxybenzoic acid ≥98.0% (by GC, titration analysis)
2-Chloro-4-methoxybenzoic acid
Formula:
C₈H₇ClO₃ |
MDL Number:
MFCD00085943 CAS Number: 21971-21-1 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 210.0 to 214.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...