Phenyl propargyl ether ≥98.0% (by GC)

Supplier: TCI America
Warning

P2222-25G P2222-5G
TCP2222-25GEA 579.86 CAD
TCP2222-25G TCP2222-5G
Phenyl propargyl ether ≥98.0% (by GC)
Phenyl propargyl ether
Formula: C₉H₈O
Boiling Pt: 83°C
Flash Pt: 76°C
Storage Temperature: Refrigerator
MDL Number: MFCD00048107
CAS Number: 13610-02-1
EINECS: 237-095-5

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR