1H,1H,12H,12H-Eicosafluoro-1,12-dodecandiol ≥96.0% (by GC)
Supplier: TCI America
Synonyms:
1H,1H,12H,12H-Perfluoro-1,12-dodecanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Eicosafluoro-1,12-dodecandiol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-Icosafluorododecane-1,12-diol, 1H,1H,12H,12H-Icosafluoro-1,12-dodecanediol, 1H,1H,12H,12H-Perfluoro-1,12-dodecandiol
I0994-1G
I0994-5G
76219-992EA
88.48
CAD
76219-992
76220-174
1H,1H,12H,12H-Eicosafluoro-1,12-dodecandiol ≥96.0% (by GC)
1H,1H,12H,12H-Eicosafluoro-1,12-dodecandiol
Formula:
C₁₂H₆F₂₀O₂ |
MDL Number:
MFCD00236625 CAS Number: 183162-43-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 165.0 to 169.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...