3'-Chloro-4'-hydroxyacetophenone ≥98.0% (by GC)

Supplier: TCI America
Warning

C3076-1G C3076-5G
TCC3076-1GEA 56.88 CAD
TCC3076-1G TCC3076-5G
3'-Chloro-4'-hydroxyacetophenone ≥98.0% (by GC)
3'-Chloro-4'-hydroxyacetophenone
Formula: C₈H₇ClO₂
MDL Number: MFCD00663621
CAS Number: 2892-29-7

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 101.0 to 105.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR