4-Iodo-3-nitrobenzoic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

I0980-1G I0980-5G
TCI0980-1GEA 56.88 CAD
TCI0980-1G TCI0980-5G
4-Iodo-3-nitrobenzoic acid ≥97.0% (by GC, titration analysis)
4-Iodo-3-nitrobenzoic acid
Formula: C₇H₄INO₄
MDL Number: MFCD00502399
CAS Number: 35674-27-2

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 210.0 to 214.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR