2-(Trifluoromethyl)benzenesulfonamide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

T3368-25G T3368-5G
76220-584EA 254.38 CAD
76220-584 76220-586
2-(Trifluoromethyl)benzenesulfonamide ≥98.0% (by HPLC, total nitrogen)
2-(Trifluoromethyl)benzenesulphonamide
Formula: C₇H₆F₃NO₂S
MDL Number: MFCD00042420
CAS Number: 1869-24-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 189.0 to 193.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR