2,4-Difluorobenzenesulfonamide ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
D5161-25G
D5161-5G
76219-196EA
330.22
CAD
76219-196
76219-198
2,4-Difluorobenzenesulfonamide ≥98.0% (by GC)
2,4-Difluorobenzenesulphonamide
Formula:
C₆H₅F₂NO₂S |
MDL Number:
MFCD01320743 CAS Number: 13656-60-5 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 154.0 to 158.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...