2-Fluoro-4-iodo-5-methylpyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-Fluoro-4-iodo-5-picoline

F1035-1G F1035-5G
TCF1035-1GEA 71.1 CAD
TCF1035-1G TCF1035-5G
2-Fluoro-4-iodo-5-methylpyridine ≥98.0% (by GC)
2-Fluoro-4-iodo-5-methylpyridine
Formula: C₆H₅FIN
MW: 237.02 g/mol
MDL Number: MFCD03095297
CAS Number: 153034-94-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 42.0 to 46.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR