4'-Fluoro-3'-methylacetophenone ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
F1097-1G
F1097-5G
TCF1097-1GEA
45.82
CAD
TCF1097-1G
TCF1097-5G
4'-Fluoro-3'-methylacetophenone ≥98.0% (by GC)
4'-Fluoro-3'-methylacetophenone
Formula:
C₉H₉FO Boiling Pt: 215°C |
MDL Number:
MFCD04972879 CAS Number: 369-32-4 |
Specification Test Results
Appearance | White to Almost white clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...