2-Fluoro-4-(trifluoromethyl)pyridine ≥98.0% (by GC)
Supplier: TCI America
|
Danger
|
F0995-1G
F0995-5G
TCF0995-1GEA
86.9
CAD
TCF0995-1G
TCF0995-5G
2-Fluoro-4-(trifluoromethyl)pyridine ≥98.0% (by GC)
2-Fluoro-4-(trifluoromethyl)pyridine
Formula:
C₆H₃F₄N Flash Pt: 23°C |
MDL Number:
MFCD09879262 CAS Number: 118078-66-3 UN: 1992 ADR: 3,III |
Specification Test Results
Appearance | White to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...