3,5-Bis(trifluoromethyl)-1H-pyrazole ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B4762-1G
B4762-5G
TCB4762-1GEA
75.84
CAD
TCB4762-1G
TCB4762-5G
3,5-Bis(trifluoromethyl)-1H-pyrazole ≥98.0% (by GC)
3,5-Bis(trifluoromethyl)-1H-pyrazole
Formula:
C₅H₂F₆N₂ Boiling Pt: 147°C |
MDL Number:
MFCD00153672 CAS Number: 14704-41-7 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 83.0 to 87.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...