3',4'-Dimethoxyflavone ≥98.0% (by GC)

Supplier: TCI America

D4794-1G D4794-200MG
TCD4794-1GEA 516.66 CAD
TCD4794-1G TCD4794-200MG
3',4'-Dimethoxyflavone ≥98.0% (by GC)
3',4'-Dimethoxyflavone
Formula: C₁₇H₁₄O₄
MDL Number: MFCD00143009
CAS Number: 4143-62-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 151.0 to 155.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR