3-Hydroxy-6-methoxyflavone ≥98.0% (by GC)

Supplier: TCI America

H1522-1G H1522-5G
TCH1522-1GEA 142.2 CAD
TCH1522-1G TCH1522-5G
3-Hydroxy-6-methoxyflavone ≥98.0% (by GC)
3-Hydroxy-6-methoxyflavone
Formula: C₁₆H₁₂O₄
MDL Number: MFCD00017639
CAS Number: 93176-00-2

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 206.0 to 210.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR