2'-Hydroxy-5'-methyl-3'-nitroacetophenone ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
H1479-25G
H1479-5G
TCH1479-25GEA
260.7
CAD
TCH1479-25G
TCH1479-5G
2'-Hydroxy-5'-methyl-3'-nitroacetophenone ≥98.0% (by GC, titration analysis)
2'-Hydroxy-5'-methyl-3'-nitroacetophenone
Formula:
C₉H₉NO₄ |
MDL Number:
MFCD00192216 CAS Number: 66108-30-3 |
Specification Test Results
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 132.0 to 136.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...