6-Methylflavone ≥98.0% (by GC)

Supplier: TCI America

M2766-1G M2766-5G
TCM2766-1GEA 115.34 CAD
TCM2766-1G TCM2766-5G
6-Methylflavone ≥98.0% (by GC)
6-Methylflavone
Formula: C₁₆H₁₂O₂
Boiling Pt: 235°C
MDL Number: MFCD00017461
CAS Number: 29976-75-8

Order Now

Specification Test Results

Appearance White to Green to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 119.0 to 123.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR