4-Methylbenzamidine hydrochloride ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
|
Warning
|
M2714-1G
M2714-5G
TCM2714-1GEA
75.84
CAD
TCM2714-1G
TCM2714-5G
4-Methylbenzamidine hydrochloride ≥98.0% (by HPLC, total nitrogen)
4-Methylbenzamidine hydrochloride
Formula:
C₈H₁₁ClN₂ |
MDL Number:
MFCD00040183 CAS Number: 6326-27-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 213.0 to 217.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...