2-Chloro-4-iodo-3-methylpyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

C3368-1G C3368-5G
TCC3368-1GEA 129.56 CAD
TCC3368-1G TCC3368-5G
2-Chloro-4-iodo-3-methylpyridine ≥98.0% (by GC)
2-Chloro-4-iodo-3-methylpyridine
Formula: C₆H₅ClIN
MDL Number: MFCD03095293
CAS Number: 153034-88-9

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 104.0 to 108.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR