1,2-Dichloro-4-fluoro-5-nitrobenzene ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
D4694-25G
D4694-5G
TCD4694-25GEA
477.16
CAD
TCD4694-25G
TCD4694-5G
1,2-Dichloro-4-fluoro-5-nitrobenzene ≥98.0% (by GC)
1,2-Dichloro-4-fluoro-5-nitrobenzene
Formula:
C₆H₂Cl₂FNO₂ Boiling Pt: 247°C Melting Pt: 17°C |
MDL Number:
MFCD00075330 CAS Number: 2339-78-8 |
Specification Test Results
Appearance | White or Colorles to Yellow to Orange powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...