5-Chloro-4-fluoro-2-nitroaniline ≥98.0% (by GC)

Supplier: TCI America
Warning

C3141-1G C3141-5G
TCC3141-1GEA 72.68 CAD
TCC3141-1G TCC3141-5G
5-Chloro-4-fluoro-2-nitroaniline ≥98.0% (by GC)
5-Chloro-4-fluoro-2-nitroaniline
Formula: C₆H₄ClFN₂O₂
MDL Number: MFCD00052698
CAS Number: 104222-34-6

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 140.0 to 144.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR