4-Chloro-3-(trifluoromethyl)benzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

C2803-1G C2803-5G
TCC2803-1GEA 26.86 CAD
TCC2803-1G TCC2803-5G
4-Chloro-3-(trifluoromethyl)benzoic acid ≥98.0% (by GC, titration analysis)
4-Chloro-3-(trifluoromethyl)benzoic acid
Formula: C₈H₄ClF₃O₂
MDL Number: MFCD01631468
CAS Number: 1737-36-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 160.0 to 165.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR