2'-Chloro-5'-(trifluoromethyl)acetophenone ≥97.0% (by GC)

Supplier: TCI America
Warning

C3336-1G C3336-5G
TCC3336-1GEA 64.78 CAD
TCC3336-1G TCC3336-5G
2'-Chloro-5'-(trifluoromethyl)acetophenone ≥97.0% (by GC)
2'-Chloro-5'-(trifluoromethyl)acetophenone
Formula: C₉H₆ClF₃O
MW: 222.59 g/mol
Flash Pt: 91°C
MDL Number: MFCD00045092
CAS Number: 71648-45-8

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR