2'-Chloro-5'-fluoroacetophenone ≥98.0% (by GC)

Supplier: TCI America
Warning

C3297-1G C3297-5G
TCC3297-1GEA 66.36 CAD
TCC3297-1G TCC3297-5G
2'-Chloro-5'-fluoroacetophenone ≥98.0% (by GC)
2'-Chloro-5'-fluoroacetophenone
Formula: C₈H₆ClFO
Boiling Pt: 82°C
MDL Number: MFCD00042574
CAS Number: 2965-16-4

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR