4-Chlorobenzamidine hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

C3090-25G C3090-5G
TCC3090-25GEA 312.84 CAD
TCC3090-25G TCC3090-5G
4-Chlorobenzamidine hydrochloride ≥98.0% (by HPLC, titration analysis)
4-Chlorobenzamidine hydrochloride
Formula: C₇H₈Cl₂N₂
MDL Number: MFCD00126401
CAS Number: 14401-51-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 244.0 to 246.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR