3-Chlorosalicylaldehyde ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

C3166-1G C3166-5G
TCC3166-1GEA 67.78 CAD
TCC3166-1G TCC3166-5G
3-Chlorosalicylaldehyde ≥98.0% (by GC, titration analysis)
3-Chlorosalicylaldehyde
Formula: C₇H₅ClO₂
Boiling Pt: 63°C
MDL Number: MFCD04973581
CAS Number: 1927-94-2

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 53.0 to 57.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR