2,5-Dibromo-3-nitropyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

D4702-1G D4702-5G
TCD4702-1GEA 31.6 CAD
TCD4702-1G TCD4702-5G
2,5-Dibromo-3-nitropyridine ≥98.0% (by GC)
2,5-Dibromo-3-nitropyridine
Formula: C₅H₂Br₂N₂O₂
MDL Number: MFCD09266223
CAS Number: 15862-37-0

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 92.0 to 96.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR