2-Bromo-5-nitro-4-picoline ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
2-Bromo-4-methyl-5-nitropyridine
B5062-25G
B5062-5G
TCB5062-25GEA
883.22
CAD
TCB5062-25G
TCB5062-5G
2-Bromo-5-nitro-4-picoline ≥98.0% (by GC)
2-Bromo-5-nitro-4-picoline
Formula:
C₆H₅BrN₂O₂ MW: 217.02 |
MDL Number:
MFCD03095066 CAS Number: 23056-47-5 |
Specification Test Results
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 63.0 to 67.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...