3-Bromo-2-fluorobenzaldehyde ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B5363-25G
B5363-5G
TCB5363-25GEA
312.84
CAD
TCB5363-25G
TCB5363-5G
3-Bromo-2-fluorobenzaldehyde ≥98.0% (by GC)
3-Bromo-2-fluorobenzaldehyde
Formula:
C₇H₄BrFO |
MDL Number:
MFCD09027089 CAS Number: 149947-15-9 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 41.0 to 45.0 deg-C |
Acid Value | max. 3.0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...