3-Bromo-5-(trifluoromethyl)benzaldehyde ≥98.0% (by GC)

Supplier: TCI America
Warning

B4974-1G B4974-5G
TCB4974-1GEA 37.92 CAD
TCB4974-1G TCB4974-5G
3-Bromo-5-(trifluoromethyl)benzaldehyde ≥98.0% (by GC)
3-Bromo-5-(trifluoromethyl)benzaldehyde
Formula: C₈H₄BrF₃O
MDL Number: MFCD08059506
CAS Number: 477535-41-4

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR