3-Bromo-6-fluoro-2-methylpyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 5-Bromo-2-fluoro-6-methylpyridine, 5-Bromo-2-fluoro-6-picoline

B5135-25G B5135-5G
TCB5135-25GEA 254.38 CAD
TCB5135-25G TCB5135-5G
3-Bromo-6-fluoro-2-methylpyridine ≥98.0% (by GC)
3-Bromo-6-fluoro-2-methylpyridine
Formula: C₆H₅BrFN
MW: 190.01
MDL Number: MFCD03095092
CAS Number: 375368-83-5

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR