4-Bromobutyl acetate ≥97.0% (by GC)
Supplier: TCI America
B4958-25G
B4958-5G
TCB4958-25GEA
123.24
CAD
TCB4958-25G
TCB4958-5G
4-Bromobutyl acetate ≥97.0% (by GC)
4-Bromobutyl acetate
Formula:
C₆H₁₁BrO₂ Boiling Pt: 211°C |
MDL Number:
MFCD00000263 CAS Number: 4753-59-7 EINECS: 225-277-7 |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...