3-Bromo-5-chloro-2-methoxypyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

B5165-1G B5165-5G
TCB5165-1GEA 158 CAD
TCB5165-1G TCB5165-5G
3-Bromo-5-chloro-2-methoxypyridine ≥98.0% (by GC)
3-Bromo-5-chloro-2-methoxypyridine
Formula: C₆H₅BrClNO
MDL Number: MFCD08059322
CAS Number: 102830-75-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 49.0 to 53.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR