2-Bromo-3-chloro-5-(trifluoromethyl)pyridine ≥96.0% (by GC)
Supplier: TCI America
|
Warning
|
B4603-1G
B4603-5G
TCB4603-1GEA
61.15
CAD
TCB4603-1G
TCB4603-5G
2-Bromo-3-chloro-5-(trifluoromethyl)pyridine ≥96.0% (by GC)
2-Bromo-3-chloro-5-(trifluoromethyl)pyridine
Formula:
C₆H₂BrClF₃N |
MDL Number:
MFCD00153072 CAS Number: 75806-84-7 |
Specification Test Results
Appearance | White to Almost white clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...