5-Bromo-2-chloro-3-fluoropyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

B5089-25G B5089-5G
TCB5089-25GEA 368.14 CAD
TCB5089-25G TCB5089-5G
5-Bromo-2-chloro-3-fluoropyridine ≥98.0% (by GC)
5-Bromo-2-chloro-3-fluoropyridine
Formula: C₅H₂BrClFN
Storage Temperature: Refrigerator
MDL Number: MFCD03411574
CAS Number: 831203-13-5

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Melting point 50.0 to 54.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR