2-Bromobenzimidazole ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
B4838-1G
B4838-5G
TCB4838-1GEA
107.44
CAD
TCB4838-1G
TCB4838-5G
2-Bromobenzimidazole ≥98.0% (by GC, titration analysis)
2-Bromobenzimidazole
Formula:
C₇H₅BrN₂ Melting Pt: 192°C |
MDL Number:
MFCD00160009 CAS Number: 54624-57-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...