3-Amino-2-bromo-5-chloropyridine ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
2-Bromo-5-chloro-3-pyridylamine
A2973-25G
A2973-5G
TCA2973-25GEA
225.94
CAD
TCA2973-25G
TCA2973-5G
3-Amino-2-bromo-5-chloropyridine ≥98.0% (by GC)
3-Amino-2-bromo-5-chloropyridine
Formula:
C₅H₄BrClN₂ MW: 207.46 g/mol |
MDL Number:
MFCD07368976 CAS Number: 90902-83-3 |
Specification Test Results
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 142.0 to 146.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...