Dextrin

Supplier: TCI America

Synonyms: Dextrin

D4657-100G D4657-500G
TCD4657-100GEA 35.38 CAD
TCD4657-100G TCD4657-500G
Dextrin
Dextrin
Formula: (C₆H₁₂O₆)n
MW: 180.16 g/mol
MDL Number: MFCD00081554
CAS Number: 9004-53-9
EINECS: 232-675-4
Merck Index: 14,02953

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Drying loss max. 10.0 %
Ignition residue (Sulfate) max. 0.5 %
Reducing suger max. 5.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR