4'-Bromo-2'-hydroxyacetophenone ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 1-(4-Bromo-2-hydroxyphenyl)ethanone, 1-(4-Bromo-2-hydroxyphenyl)ethan-1-one

B5382-1G B5382-5G
TCB5382-1GEA 36.34 CAD
TCB5382-1G TCB5382-5G
4'-Bromo-2'-hydroxyacetophenone ≥98.0% (by GC, titration analysis)
4'-Bromo-2'-hydroxyacetophenone
Formula: C₈H₇BrO₂
MW: 215.05
Boiling Pt: 83°C
MDL Number: MFCD03428533
CAS Number: 30186-18-6

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 40.0 to 44.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR