3-Aminophthalic acid ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Warning

A1516-1G A1516-5G
TCA1516-1GEA 74.26 CAD
TCA1516-1G TCA1516-5G
3-Aminophthalic acid ≥95.0% (by titrimetric analysis)
3-Aminophthalic acid
Formula: C₈H₇NO₄
CAS Number: 5434-20-8
EINECS: 226-595-9

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 93.0 area%
Purity (Neutralization titration) min. 95.0 %
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR