3-Fluoro-5-(trifluoromethyl)benzonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

F0994-25G F0994-5G
TCF0994-25GEA 666.76 CAD
TCF0994-25G TCF0994-5G
3-Fluoro-5-(trifluoromethyl)benzonitrile ≥98.0% (by GC)
3-Fluoro-5-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₄N
Flash Pt: 65°C
MDL Number: MFCD00061282
CAS Number: 149793-69-1
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR