3-Fluoro-4-(trifluoromethyl)benzonitrile ≥99.0% (by GC)

Supplier: TCI America
Danger

F1019-1G F1019-5G
TCF1019-1GEA 41.08 CAD
TCF1019-1G TCF1019-5G
3-Fluoro-4-(trifluoromethyl)benzonitrile ≥99.0% (by GC)
3-Fluoro-4-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₄N
MDL Number: MFCD00236280
CAS Number: 231953-38-1
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 46.0 to 50.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR