1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol ≥98.0% (by GC)

Supplier: TCI America
Warning

H1349-25G H1349-5G
76220-120EA 584.6 CAD
76220-120 76220-122
1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol ≥98.0% (by GC)
1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol
Formula: C₄H₄F₆O
Boiling Pt: 62°C
MDL Number: MFCD00039259
CAS Number: 1515-14-6
EINECS: 216-155-4

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR