1,3-Dimethylimidazolium methyl sulfate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

Synonyms: 1,3-Dimethyl-1H-imidazol-3-ium methyl sulphate, 1,3-Dimethyl-1H-imidazol-3-ium methyl sulfate

D5348-25G D5348-5G
76219-928EA 363.4 CAD
76219-928 76219-930
1,3-Dimethylimidazolium methyl sulfate ≥98.0% (by HPLC, total nitrogen)
1,3-Dimethylimidazolium methyl sulphate
Formula: C₆H₁₂N₂O₄S
MDL Number: MFCD03453276
CAS Number: 97345-90-9

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR