2-Acetylphenothiazine ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Methyl phenothiazin-2-yl ketone, 1-(10H-Phenothiazin-2-yl)ethanone, ML171

A2815-25G A2815-5G
TCA2815-25GEA 905.34 CAD
TCA2815-25G TCA2815-5G
2-Acetylphenothiazine ≥98.0% (by GC)
2-Acetylphenothiazine
Formula: C₁₄H₁₁NOS
Storage Temperature: Refrigerator
MDL Number: MFCD00005017
CAS Number: 6631-94-3
EINECS: 229-626-4

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 190.0 to 194.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR