2-Bromo-3-pyridineboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

B5170-1G B5170-5G
TCB5170-1GEA 99.54 CAD
TCB5170-1G TCB5170-5G
2-Bromo-3-pyridineboronic acid (contains varying amounts of Anhydride)
2-Bromo-3-pyridineboronic acid
Formula: C₅H₅BBrNO₂
MDL Number: MFCD04113805
CAS Number: 452972-08-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) lt. 98.0 area%
Purity (Neutralization titration) 97.0 to 110.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR