2-Fluoro-6-methoxybenzonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

F0969-25G F0969-5G
TCF0969-25GEA 279.34 CAD
TCF0969-25G TCF0969-5G
2-Fluoro-6-methoxybenzonitrile ≥98.0% (by GC)
2-Fluoro-6-methoxybenzonitrile
Formula: C₈H₆FNO
MDL Number: MFCD00042291
CAS Number: 94088-46-7
EINECS: 302-047-5
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 102.0 to 106.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR